5-Bromo-3-ethynylpyridin-2-ylamine 97%

Supplier: Ambeed
Warning

A124680-1G A124680-250MG
76781-930EA 254.62 USD
76781-930 76781-932
5-Bromo-3-ethynylpyridin-2-ylamine 97%
5-Bromo-3-ethynylpyridin-2-ylamine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD10566045
CAS Number: 1210838-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR