4-Amino-3-iodobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Cyano-2-iodoaniline
A124517-100G
A124517-10G
A124517-1G
A124517-25G
A124517-5G
76781-918EA
233.4
USD
76781-918
76781-920
76781-922
76781-924
76781-926
4-Amino-3-iodobenzonitrile 98%
4-Amino-3-iodobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₅IN₂ MW: 244.03 g/mol |
MDL Number:
MFCD04039964 CAS Number: 33348-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...