Zanamivir 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S)-3-acetamido-4-(diaminomethylideneamino)-2-[(1R,2R)-1,2,3-trihydroxypropyl]-3,4-dihydro-2H-pyran-6-carboxylic acid, 4-Guanidino-Neu5Ac2en, (2R,3R,4S)-3-Acetamido-4-guanidino-2-((1R,2R)-1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid, 4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid

A124461-10MG A124461-1MG A124461-5MG A124461-1G A124461-250MG A124461-50MG
76781-912EA 23.34 USD
76781-912 76781-914 76781-916 77566-382 77666-776 77666-778
Zanamivir 98%
Zanamivir

Precaución: For research use only.

Formula: C₁₂H₂₀N₄O₇
MW: 332.31 g/mol
MDL Number: MFCD00866966
CAS Number: 139110-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR