3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

A124362-1G A124362-25G A124362-5G
76781-906EA 10.61 USD
76781-906 76781-908 76781-910
3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%
3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₁H₂₄F₂
MW: 314.42 g/mol
MDL Number: MFCD09838999
CAS Number: 85312-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR