4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A124267-10G A124267-250MG A124267-5G
76781-892EA 299 USD
76781-892 76781-894 77871-559
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 95%
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MDL Number: MFCD12406103
CAS Number: 1086064-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR