Xylometazoline hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: 2-(4-(tert-Butyl)-2,6-dimethylbenzyl)-4,5-dihydro-1H-imidazole hydrochloride, 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline Hydrochloride

A124255-100MG A124255-1G A124255-25G A124255-5G
76781-884EA 5.31 USD
76781-884 76781-886 76781-888 76781-890
Xylometazoline hydrochloride 97%
Xylometazoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂
MW: 280.84 g/mol
MDL Number: MFCD00238707
CAS Number: 1218-35-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR