1-Phenylethyl acetate 98%

Supplier: Ambeed
Warning

A124116-1000G A124116-100G A124116-10G A124116-25G A124116-500G
76781-874EA 152 USD
76781-874 76781-876 76781-878 76781-880 76781-882
1-Phenylethyl acetate 98%
1-Phenylethyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00008858
CAS Number: 93-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR