3-Bromo-2-nitroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-methoxy-2-nitrobenzene

A123639-1G A123639-250MG A123639-5G A123639-25G
76781-864EA 14 USD
76781-864 76781-866 76781-868 77871-556
3-Bromo-2-nitroanisole 97%
3-Bromo-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD11101044
CAS Number: 500298-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR