2-(2-(2-Aminoethoxy)ethoxy)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A122829-100MG A122829-1G A122829-250MG A122829-5G
76781-834EA 7.43 USD
76781-834 76781-836 76781-838 76781-840
2-(2-(2-Aminoethoxy)ethoxy)acetic acid hydrochloride 97%
2-(2-(2-Aminoethoxy)ethoxy)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₄
MW: 199.63 g/mol
MDL Number: MFCD11865374
CAS Number: 134979-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR