Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloro-1H-1,2,4-triazole-3-carboxylate

A122571-1G A122571-250MG
76781-830EA 127.31 USD
76781-830 76781-832
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate 95%
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O₂
MW: 175.5700 g/mol
MDL Number: MFCD11040186
CAS Number: 774608-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR