2,2,6,6-Tetramethylpiperidinooxy (TEMPO) 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
TEMPO, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethylpiperidine 1-oxyl
A121835-10G
A121835-25G
76781-806EA
6.37
USD
76781-806
76781-808
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) 97%
2,2,6,6-Tetramethylpiperidinooxy (TEMPO)
Precaución: for Research Use Only
Formula:
C₉H₁₈NO MW: 156.25 g/mol |
MDL Number:
MFCD00009599 CAS Number: 2564-83-2 UN: 3263 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...