3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A121674-1G A121674-1MG A121674-250MG A121674-5G A121674-5MG A121674-100MG A121674-25G
76781-788EA 126 USD
76781-788 76781-790 76781-792 76781-794 76781-796 77871-538 77871-539
3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid 98%
3-(6-{[1-(2,2-Difluorobenzo[1,3]dioxol-5-yl)cyclopropanecarbonyl]amino}-3-methylpyridin-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₈F₂N₂O₅
MDL Number: MFCD16659051
CAS Number: 936727-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR