3-Fluoro-2-nitroaniline 98%
Supplier: Ambeed
|
Warning
|
A121494-100G
A121494-10G
A121494-1G
A121494-25G
A121494-500G
A121494-5G
76781-776EA
186
USD
76781-776
76781-778
76781-780
76781-782
76781-784
76781-786
3-Fluoro-2-nitroaniline 98%
3-Fluoro-2-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₅FN₂O₂ MW: 156.1145 g/mol |
MDL Number:
MFCD07368750 CAS Number: 567-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...