ML-7 hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hexahydro-1-[(5-iodo-1-naphthalenyl)sulphonyl]-1H-1,4-diazepi-ne hydrochloride, 1-((5-Iodonaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride, ML-7 HCl
A121204-10MG
A121204-1MG
A121204-50MG
A121204-5MG
76781-758EA
93.36
USD
76781-758
76781-760
76781-762
76781-764
ML-7 hydrochloride 98%
ML-7 hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₁₇IN₂O₂S.HCl MW: 452.74 g/mol |
MDL Number:
MFCD00065524 CAS Number: 110448-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...