tert-Butyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A120642-250MG A120642-5G A120642-100MG A120642-1G A120642-25G
76781-736EA 18 USD
76781-736 76781-738 77666-728 77666-730 77871-528
tert-Butyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate 97%
tert-Butyl (cis-4-(hydroxymethyl)cyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₃
MDL Number: MFCD03844606
CAS Number: 223131-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR