N-Boc-S-benzyl-D-cysteine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-Cys(Bzl)-OH
A120579-100G
A120579-10G
A120579-1G
A120579-25G
A120579-5G
76781-712EA
433.91
USD
76781-712
76781-714
76781-716
76781-718
76781-720
N-Boc-S-benzyl-D-cysteine 98%
N-Boc-S-benzyl-D-cysteine
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₄S MW: 311.396 g/mol |
MDL Number:
MFCD00038253 CAS Number: 102830-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...