2-(4-Methoxyphenyl)-1,3-thiazole-5-carbaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4-Methoxyphenyl)thiazole-5-carbaldehyde
A120208-100MG
A120208-250MG
76781-692EA
40.32
USD
76781-692
76781-694
2-(4-Methoxyphenyl)-1,3-thiazole-5-carbaldehyde 98%
2-(4-Methoxyphenyl)-1,3-thiazole-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₂S MW: 219.26 g/mol |
MDL Number:
MFCD05864660 CAS Number: 914348-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...