7-Bromo-1H-imidazo[4,5-c]pyridine 97%

Supplier: Ambeed
Warning

A119791-100MG A119791-1G A119791-250MG A119791-5G
76781-668EA 49.87 USD
76781-668 76781-670 76781-672 76781-674
7-Bromo-1H-imidazo[4,5-c]pyridine 97%
7-Bromo-1H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD09834585
CAS Number: 90993-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR