Benzylhydrazine monohydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: Benzylhydrazine hydrochloride

A119625-1G A119625-5G A119625-10G A119625-100G
76781-650EA 7.43 USD
76781-650 76781-652 77549-402 77666-708
Benzylhydrazine monohydrochloride 95%
Benzylhydrazine monohydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 158.63 g/mol
MDL Number: MFCD01722685
CAS Number: 1073-62-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR