Trimethylphosphine oxide 98%

Supplier: Ambeed
Warning

Synonyms: Trimethylphosphineoxide

A119201-100G A119201-1G A119201-25G A119201-5G
76781-624EA 1262.48 USD
76781-624 76781-626 76781-628 76781-630
Trimethylphosphine oxide 98%
Trimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₃H₉OP
MW: 92.08 g/mol
MDL Number: MFCD00013910
CAS Number: 676-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR