N-Boc-3-methyl-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(m-tolyl)propanoic acid

A118924-1G A118924-250MG A118924-5G
76781-596EA 31 USD
76781-596 76781-598 76781-600
N-Boc-3-methyl-D-phenylalanine 98%
N-Boc-3-methyl-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD01862943
CAS Number: 114873-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR