Tetrabutylammonium chloride hydrate 97%

Supplier: Ambeed
Warning

Synonyms: Tetrabutylammonium chloride monohydrate, TBAC

A118906-100G A118906-10G A118906-25G A118906-500G
76781-588EA 20.16 USD
76781-588 76781-590 76781-592 76781-594
Tetrabutylammonium chloride hydrate 97%
Tetrabutylammonium chloride hydrate

Precaución: for Research Use Only

Formula: C₁₆H₃₆ClN·H₂O
MW: 295.94 g/mol
MDL Number: MFCD00149970
CAS Number: 88641-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR