tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(pyrimidin-2-yloxy)piperidine-1-carboxylate

A118719-100MG A118719-1G A118719-250MG A118719-25G A118719-5G
76781-578EA 16.98 USD
76781-578 76781-580 76781-582 76781-584 76781-586
tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate 97%
tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₃O₃
MW: 279.3400 g/mol
MDL Number: MFCD09028557
CAS Number: 412293-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR