4-(Dibenzylamino)cyclohexanone 95%

Supplier: Ambeed
Warning

A118656-1G A118656-250MG A118656-25G A118656-5G
76781-568EA 33.99 USD
76781-568 76781-570 76781-572 76781-574
4-(Dibenzylamino)cyclohexanone 95%
4-(Dibenzylamino)cyclohexanone

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO
MDL Number: MFCD16659682
CAS Number: 149506-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR