4-Cyano-3,5-difluoropyridine 98%

Supplier: Ambeed
Warning

A118346-1G A118346-5G
76781-540EA 14 USD
76781-540 76781-542
4-Cyano-3,5-difluoropyridine 98%
4-Cyano-3,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₆H₂F₂N₂
MDL Number: MFCD13176565
CAS Number: 1214377-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR