4-Benzyloxy-2-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A118309-1G A118309-25G A118309-5G
76781-534EA 21.99 USD
76781-534 76781-536 76781-538
4-Benzyloxy-2-methylbenzaldehyde 95%
4-Benzyloxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MDL Number: MFCD01317809
CAS Number: 101093-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR