4-Fluoro-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A118062-100MG A118062-1G A118062-250MG A118062-25G A118062-5G A118062-100G A118062-10G
76781-514EA 5 USD
76781-514 76781-516 76781-518 76781-520 76781-522 77871-507 77871-508
4-Fluoro-3-methoxyphenylboronic acid 98%
4-Fluoro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MDL Number: MFCD08056358
CAS Number: 854778-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR