1,6-Naphthyridin-5(6H)-one 97%

Supplier: Ambeed
Warning

A118029-100MG A118029-250MG
76781-508EA 22 USD
76781-508 76781-510
1,6-Naphthyridin-5(6H)-one 97%
1,6-Naphthyridin-5(6H)-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD08567947
CAS Number: 23616-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR