Salicylaldehyde azine 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2-hydroxybenzaldehyde) hydrazone, N,N'-Bis(salicylidene)hydrazine, Salazine, Salicylaldazine, 2,2'-(Hydrazine-1,2-diylidenebis(methanylylidene))diphenol

A117926-100G A117926-25G A117926-5G
76781-494EA 72.15 USD
76781-494 76781-496 76781-498
Salicylaldehyde azine 97%
Salicylaldehyde azine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
MDL Number: MFCD00043496
CAS Number: 959-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR