Thiamine (Vitamin B1) 98%

Supplier: Ambeed
Warning

Synonyms: Thiamine monochloride, Aneurine, Vitamin B1, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride

A117613-10G A117613-25G A117613-5G A117613-500G A117613-1000G A117613-100G
76781-488EA 18 USD
76781-488 76781-490 76781-492 77599-698 77871-504 77871-505
Thiamine (Vitamin B1) 98%
Thiamine (Vitamin B1)

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClN₄OS
MDL Number: MFCD00044586
CAS Number: 59-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR