Dimethyl-2,6-naphthalenedicarboxylate 99%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2,6-naphthalenedicarboxylate, 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester

A117385-100G A117385-10G A117385-1G A117385-25G A117385-5G
76781-472EA 214.31 USD
76781-472 76781-474 76781-476 76781-478 76781-480
Dimethyl-2,6-naphthalenedicarboxylate 99%
Dimethyl-2,6-naphthalenedicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
MDL Number: MFCD00004100
CAS Number: 840-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR