trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
Supplier: Ambeed
|
Warning
|
A117087-100G
A117087-10G
A117087-25G
A117087-5G
A117087-500G
76781-456EA
66
USD
76781-456
76781-458
76781-460
76781-462
77666-682
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₂O₈·1H₂O |
MDL Number:
MFCD00066429 CAS Number: 125572-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...