2-Amino-5-bromobenzo[d]thiazole-3-oxide 95+%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromobenzo[d]thiazole 3-oxide

A117045-25G A117045-5G
76781-452EA 1003.62 USD
76781-452 76781-454
2-Amino-5-bromobenzo[d]thiazole-3-oxide 95+%
2-Amino-5-bromobenzo[d]thiazole-3-oxide

Precaución: for Research Use Only

Formula: C₇H₅BrN₂OS
MW: 245.1 g/mol
MDL Number: MFCD12406142
CAS Number: 1216671-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR