trans-Cinnamaldehyde 98%

Supplier: Ambeed
Warning

A116977-1000G A116977-100G A116977-25G A116977-500G
76781-444EA 385 USD
76781-444 76781-446 76781-448 76781-450
trans-Cinnamaldehyde 98%
trans-Cinnamaldehyde

Precaución: for Research Use Only

Formula: C₉H₈O
MDL Number: MFCD00007000
CAS Number: 14371-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR