Tetrakis(2-methoxyethyl) orthosilicate 95+%

Supplier: Ambeed
Warning

A116933-100G A116933-25G A116933-500G
76781-430EA 47.75 USD
76781-430 76781-432 76781-434
Tetrakis(2-methoxyethyl) orthosilicate 95+%
Tetrakis(2-methoxyethyl) orthosilicate

Precaución: for Research Use Only

Formula: C₁₂H₂₈O₈Si
MW: 328.43 g/mol
MDL Number: MFCD00025907
CAS Number: 2157-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR