Darunavir 98%

Supplier: Ambeed
Warning

A116353-100MG A116353-10MG A116353-1G A116353-1MG A116353-250MG A116353-5G A116353-5MG A116353-25G
76781-404EA 23 USD
76781-404 76781-406 76781-408 76781-410 76781-412 76781-414 76781-416 77871-485
Darunavir 98%
Darunavir

Precaución: for Research Use Only

Formula: C₂₇H₃₇N₃O₇S
MDL Number: MFCD09260006
CAS Number: 206361-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR