(±)-trans-1,2-Cyclohexanediol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol
A116284-10G
A116284-5G
A116284-500G
A116284-25G
76781-400EA
22.28
USD
76781-400
76781-402
77599-680
77666-678
(±)-trans-1,2-Cyclohexanediol 95%
(±)-trans-1,2-Cyclohexanediol
Precaución: for Research Use Only
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol |
MDL Number:
MFCD00063611 CAS Number: 1460-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...