Methyl 1-(diphenylmethyl)azetidine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-benzhydrylazetidine-3-carboxylate

A116232-1G A116232-5G
76781-396EA 9.55 USD
76781-396 76781-398
Methyl 1-(diphenylmethyl)azetidine-3-carboxylate 97%
Methyl 1-(diphenylmethyl)azetidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₂
MW: 281.349 g/mol
MDL Number: MFCD03407521
CAS Number: 53871-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR