Thieno[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A115920-100MG A115920-1G A115920-250MG
76781-388EA 30.77 USD
76781-388 76781-390 76781-392
Thieno[3,2-c]pyridine 95%
Thieno[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅NS
MW: 135.1863 g/mol
MDL Number: MFCD01760493
CAS Number: 272-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR