5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A115463-1G A115463-250MG A115463-5G
76781-360EA 28 USD
76781-360 76781-362 76781-364
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid 97%
5-Amino-2-(methylthio)pyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂S
MDL Number: MFCD12407810
CAS Number: 100130-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR