3-Bromo-2-cyano-6-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-methylpicolinonitrile, 3-Bromo-6-methylpyridine-2-carbonitrile

A114873-100MG A114873-250MG A114873-5G
76781-338EA 16.98 USD
76781-338 76781-340 77566-038
3-Bromo-2-cyano-6-methylpyridine 98%
3-Bromo-2-cyano-6-methylpyridine

Precaución: For research use only.

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD08059560
CAS Number: 717843-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR