Z-Leu-ONp 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenyl ((benzyloxy)carbonyl)-L-leucinate

A114822-10G A114822-1G A114822-5G
76781-332EA 28.65 USD
76781-332 76781-334 76781-336
Z-Leu-ONp 97%
Z-Leu-ONp

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₆
MW: 386.404 g/mol
MDL Number: MFCD00024656
CAS Number: 1738-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR