2-Bromo-6-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A114752-100MG A114752-10G A114752-1G A114752-250MG A114752-25G A114752-5G
76781-316EA 6.37 USD
76781-316 76781-318 76781-320 76781-322 76781-324 77666-666
2-Bromo-6-iodobenzonitrile 95%
2-Bromo-6-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MW: 307.9139 g/mol
MDL Number: MFCD16619725
CAS Number: 1245648-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR