5-Nitro-2,3,4-trifluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,3,4-Trifluoro-5-nitrobenzoic acid

A114523-10G A114523-1G A114523-25G A114523-5G
76781-296EA 32 USD
76781-296 76781-298 76781-300 76781-302
5-Nitro-2,3,4-trifluorobenzoic acid 97%
5-Nitro-2,3,4-trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂F₃NO₄
MDL Number: MFCD09753762
CAS Number: 197520-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR