5-Bromo-6-chloropyrimidin-4-amine 98%

Supplier: Ambeed
Warning

A114298-1G A114298-250MG A114298-5G A114298-25G
76781-274EA 220.67 USD
76781-274 76781-276 76781-278 76781-280 77666-662
5-Bromo-6-chloropyrimidin-4-amine 98%
5-Bromo-6-chloropyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₄H₃BrClN₃
MDL Number: MFCD09999224
CAS Number: 663193-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR