Thiophen-2-carbothioamide 97%

Supplier: Ambeed
Warning

Synonyms: Thiophene-2-carbothioamide

A113923-10G A113923-1G A113923-5G
76781-268EA 167.63 USD
76781-268 76781-270 76781-272
Thiophen-2-carbothioamide 97%
Thiophen-2-carbothioamide

Precaución: for Research Use Only

Formula: C₅H₅NS₂
MW: 143.23 g/mol
MDL Number: MFCD00052890
CAS Number: 20300-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR