3,6-Dimethylsalicylaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-3,6-dimethylbenzaldehyde, 3,6-Dimethyl-2-hydroxybenzaldehyde

A113873-10G A113873-1G A113873-250MG A113873-5G
76781-260EA 256.74 USD
76781-260 76781-262 76781-264 76781-266
3,6-Dimethylsalicylaldehyde 95%
3,6-Dimethylsalicylaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
MDL Number: MFCD04037328
CAS Number: 1666-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR