7-Bromothieno[3,2-d]pyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A113478-1G A113478-250MG A113478-5G
76781-236EA 82.76 USD
76781-236 76781-238 76781-240
7-Bromothieno[3,2-d]pyrimidin-4(3H)-one 95%
7-Bromothieno[3,2-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₃BrN₂OS
MW: 231.0698 g/mol
MDL Number: MFCD11518998
CAS Number: 31169-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR