(trans,trans)-4-Butyl-4'-vinyl-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

A113374-10G A113374-1G A113374-5G
76781-230EA 35 USD
76781-230 76781-232 76781-234
(trans,trans)-4-Butyl-4'-vinyl-1,1'-bi(cyclohexane) 97%
(trans,trans)-4-Butyl-4'-vinyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₁₈H₃₂
MW: 248.45 g/mol
MDL Number: MFCD11053345
CAS Number: 153429-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR