3-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(3-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxycarbonylphenylboronic acid pinacol ester

A112753-10G A112753-1G A112753-250MG A112753-5G A112753-25G
76781-212EA 36.08 USD
76781-212 76781-214 76781-216 76781-218 77666-638
3-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
3-(Methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MW: 262.1093 g/mol
MDL Number: MFCD03789258
CAS Number: 480425-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR